missing translation for 'onlineSavingsMsg'
Learn More

MCOPPB trihydrochloride, Tocris Bioscience™

Potent NOP receptor agonist

Marke:  RD Systems (Bio-Techne) 37631

Weitere Details : CAS : 1028969-49-4 Netto-Gewicht : 0.01000kg

Menge. 1mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413574

  • 224.41 € / 1 Milligramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

High affinity NOP receptor agonist (pKi = 10.07 for the human NOP receptor). Exhibits anxiolytic effects with no effect on memory or locomotion. Regarded as one of the most potent, non-peptide NOP full agonists in vitro.
Spezifikation

Chemische Identifikatoren

1108147-88-1
518.008
mcoppb trihydrochloride, 1-1-1-methylcyclooctyl-4-piperidinyl-2-3r-3-piperidinyl-1h-benzimidazole trihydrochloride, mcoppb hydrochloride, mcoppb trihydrochloride/, mcoppb trihydrochloride, 1108147-88-1, r-1-1-1-methylcyclooctyl piperidin-4-yl-2-piperidin-3-yl-1h-benzo d imidazole trihydrochloride, 1-1-1-methylcyclooctyl piperidin-4-yl-2-3r-piperidin-3-yl-1,3-benzodiazole trihydrochloride
1-[1-(1-methylcyclooctyl)piperidin-4-yl]-2-[(3R)-piperidin-3-yl]benzimidazole;trihydrochloride
C26H43Cl3N4
DTIPEVOPCGEULQ-RFCADEKQSA-N
25208093
CC1(CCCCCCC1)N2CCC(CC2)N3C4=CC=CC=C4N=C3C5CCCNC5.Cl.Cl.Cl

Spezifikation

MCOPPB trihydrochloride
1108147-88-1
mcoppb trihydrochloride, 1-1-1-methylcyclooctyl-4-piperidinyl-2-3r-3-piperidinyl-1h-benzimidazole trihydrochloride, mcoppb hydrochloride, mcoppb trihydrochloride/, mcoppb trihydrochloride, 1108147-88-1, r-1-1-1-methylcyclooctyl piperidin-4-yl-2-piperidin-3-yl-1h-benzo d imidazole trihydrochloride, 1-1-1-methylcyclooctyl piperidin-4-yl-2-3r-piperidin-3-yl-1,3-benzodiazole trihydrochloride
CC1(CCCCCCC1)N2CCC(CC2)N3C4=CC=CC=C4N=C3C5CCCNC5.Cl.Cl.Cl
518.008
534.05
1mg
C26H43Cl3N4
DTIPEVOPCGEULQ-RFCADEKQSA-N
1-[1-(1-methylcyclooctyl)piperidin-4-yl]-2-[(3R)-piperidin-3-yl]benzimidazole;trihydrochloride
25208093
>99%
Videos