missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCRS1 Mouse anti-Human, Mouse, Rat, Clone: 2H10E10, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60215-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity.Spezifikation
MCRS1 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96EZ8, Q99L90 | |
Mcrs1 | |
MCRS1 Fusion Protein Ag1904 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG2a |
Western Blot | |
2H10E10 | |
Unconjugated | |
MCRS1 | |
58 kDa microspherule protein, ICP22BP, INO80 complex subunit J, INO80Q, MCRS1, MCRS2, microspherule protein 1, MSP58, P78 | |
Mouse | |
Protein A | |
RUO | |
10445, 300222, 51812 | |
-20°C | |
Liquid |