Beschreibung
MDG1 Polyclonal antibody specifically detects MDG1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Spezifikation
Spezifikation
| Antigen | MDG1 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500 - 1:2000, ELISA |
| Zusammensetzung | PBS (pH 7.3), 50% glycerol |
| Gen-Alias | DKFZp564F1862, DnaJ (Hsp40) homolog, subfamily B, member 9, dnaJ homolog subfamily B member 9, endoplasmic reticulum DnaJ homolog 4, ERdj4, Mdg-1, MDG1MSTP049, Microvascular endothelial differentiation gene 1 protein, MST049 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 24-223 of human MDG1 (NP_036460.1).,, Sequence:, SKSYYDILGVPKSASERQIKKAFHKLAMKYHPDKNKSPDAEAKFREIAEAYETLSDANRRKEYDTLGHSAFTSGKGQRGSGSSFEQSFNFNFDDLFKDFGFFGQNQNTGSKKRFENHFQTRQDGGSSRQRHHFQEFSFGGGLFDDMFEDMEKMFSFSGFDSTNQHTVQTENRFHGSSKHCRTVTQRRGNMVTTYTDCSGQ |
| Reinigungsverfahren | Affinity purified |
| Mehr anzeigen |