Learn More
MDL-28170, MedChemExpress
Product Code. 16449967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16449967 Supplier Medchem Express Supplier No. HY18236
 Restricted Item
Verfügbar ab: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress MDL-28170 (Calpain Inhibitor III) is a potent, selective and membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier following systemic administration. MDL-28170 also block γ-secretase.
MDL-28170 (Calpain Inhibitor III) is a potent, selective and membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier following systemic administration. MDL-28170 also block γ-secretase.
Specifications
Chemical Name or Material MDL-28170
Physical Form Solid
CAS 88191-84-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H26N2O4
For Use With (Application) COVID-19-immunoregulation
Synonym Calpain Inhibitor III
Solubility Information DMSO : 30 mg/mL (78.44 mM; Need ultrasonic and warming)
SMILES O=C(N[C@@H](C(C)C)C(NC(CC1=CC=CC=C1)C=O)=O)OCC2=CC=CC=C2
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 97.0%
Grade Research