missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEF2B Polyclonal Antibody

Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The myocyte enhancer factor-2 (MEF-2) family of transcription factors associated with co-repessors or co-activators to regulate development and function of T cells, neuronal cells and muscle cells. Four family members arise from alternatively spliced transcripts, termed MEF-2A, -2B, -2C and -2D. These members bind as homo- and heterodimers to the MEF-2 site in the promoter region of affected genes. Differential regulation in the expression of the four transcripts implies functional distinction for each during embryogenesis and development. The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression.
Spezifikation
Spezifikation
| Antigen | MEF2B |
| Anwendungen | Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 2.03 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gen | MEF2B |
| Gen-Zugriffsnummer | Q02080 |
| Gen-Alias | 2310045N01Rik; AI451606; BLOC-1-related complex subunit 8; Borcs8; BORCS8-MEF2B; BORCS8-MEF2B readthrough; LOC729991-MEF2B; LOC729991-MEF2B readthrough; MADS box transcription enhancer factor 2, polypeptide B (myocyte enhancer factor 2B); MEF2B; MEF2B neighbor gene protein homolog; Mef2bnb; MEF2BNB-MEF2B; MEF2BNB-MEF2B readthrough; myocyte enhancer factor 2B; myocyte-specific enhancer factor 2B; protein MEF2BNB; RIKEN cDNA 2310045N01 gene; RSRFR2; Serum response factor-like protein 2; XMEF2 |
| Gensymbole | MEF2B |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?