missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15758-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
MAP2K5 is a dual specificity protein kinase that belongs to the MAP kinase kinase family. It specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation.Spezifikation
MEK5 | |
Polyclonal | |
Unconjugated | |
MAP2K5 | |
HsT17454, MAP kinase kinase 5, MAP2K5, MAPK/ERK kinase 5, MAPKK 5, MAPKK5, MEK 5, MEK5, MKK5, PRKMK5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23938, 29568, 5607 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13163, Q62862, Q9WVS7 | |
Map2k5 | |
MEK5 Fusion Protein Ag8351 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |