missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15159-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.Spezifikation
MEN1 | |
Polyclonal | |
Unconjugated | |
MEN1 | |
MEAI, MEN1, Menin, multiple endocrine neoplasia I, SCG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4221 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00255 | |
MEN1 | |
MEN1 Fusion Protein Ag7253 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |