missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromacil in Methanol 100μ g/ml, Fisher Chemical™
Click to view available options
Menge:
1 ml
Verpackung:
Ampulle(n)
Spezifikation
Chemischer Name oder Material | Bromacil |
Konzentration | 100 μg/ml |
CAS | 314-40-9 |
Verpackung | Ampulle(n) |
Menge | 1 ml |
Summenformel | C9H13BrN2O2 |
InChI-Schlüssel | CTSLUCNDVMMDHG-UHFFFAOYSA-N |
SMILES | CCC(C)N1C(=O)C(=C(NC1=O)C)Br |
IUPAC-Name | 5-Brom-3-Butan-2-yl-6-Methyl-1 H-Pyrimidin-2,4-dion |
Molekulargewicht (g/mol) | 261.119 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'