missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metenolon-Enanthat, 1 mg/ml in Isopropanol, Lipomed
Beschreibung
Metenolon-Enanthat, 1 mg/ml in Isopropanol
Spezifikation
Spezifikation
| Chemischer Name oder Material | Primobolan |
| CAS | 303-42-4 |
| Bezeichnung der Analyten oder Bestandteile | Primobolan |
| Versandbedingung | Room Temperature |
| Summenformel | C27 H42 O3 |
| Konzentration | 1 mg/mL |
| Synonym | Metenolone enanthate, Methenolone 17-enanthate, Methenolone enanthate, NSC 64967, Primobolan Depot, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate |
| InChI-Formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCCCCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CC[C@H]4 CC(=O)C=C(C)[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC-Name | [(5S,8R,9S,10S,13S,14S,17S)-1,10,13-Trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]heptanoat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?