missing translation for 'onlineSavingsMsg'
Learn More

Meteorin Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication

N.V.

Spezifikation

Antigen Meteorin
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1 Gly22-Asp291 Accession # Q8C1Q4
Gen-ID (Entrez) 79006
Gen-Zugriffsnummer Q8C1Q4
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

Meteorin Polyclonal specifically detects Meteorin in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Spezifikation

Spezifikation

Meteorin
79006
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects mouse Meteorin in direct ELISAs and Western blots.
Unconjugated
Western Blot, Immunocytochemistry
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1 Gly22-Asp291 Accession # Q8C1Q4
Q8C1Q4
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only