Learn More
Thermo Scientific™ Methanol, anhydrous, 99.9%, packaged under Argon in resealable AcroSeal™ bottles
Product Code. 16308227
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16308227 1 L 1 L
16398217 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16308227 Supplier Thermo Scientific™ Supplier No. 047339.K2
  • ONLINE EXCLUSIVE PRICE
    74.60 €
    64.65 € / 1 Liter
    Sparen 9.95 €
    13% Rabatt