missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Methionine Aminopeptidase 1/ METAP1 Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | Methionine Aminopeptidase 1/METAP1 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Anwendungen | Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Beschreibung
Methionine Aminopeptidase 1/METAP1 Polyclonal specifically detects Methionine Aminopeptidase 1/METAP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Spezifikation
Methionine Aminopeptidase 1/METAP1 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
DKFZp781C0419, EC 3.4.11.18, KIAA0094MAP 1, MAP1, MAP1A, MetAP 1, MetAP1, MetAP1A, methionine aminopeptidase 1, methionyl aminopeptidase 1, Peptidase M 1 | |
METAP1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P53582 | |
23173 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Methionine Aminopeptidase 1/METAP1 His52-Phe386 Accession # P53582 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Methionine Aminopeptidase 1/METAP1 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse METAP1 is observed. |
For Research Use Only