missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2,3,6-tri-O-benzoyl-alpha-D-galactopyranosid, 98 %, Thermo Scientific Chemicals

Catalog Number p-7043212
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3601-36-3
C28H26O9
506.507
MFCD06300842
WXFFEILSURAFKL-SUWSLWCISA-N
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
10929259
[(2R,3S,4S,5R,6S)-4,5-Dibenzoyloxy-3-Hydroxy-6-Methoxyoxan-2-yl]methylbenzoat
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3601-36-3
C28H26O9
506.507
MFCD06300842
WXFFEILSURAFKL-SUWSLWCISA-N
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
10929259
[(2R,3S,4S,5R,6S)-4,5-Dibenzoyloxy-3-Hydroxy-6-Methoxyoxan-2-yl]methylbenzoat
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside
Schmelzpunkt 143°C
Farbe Weiß
Menge 1 g
Optische Drehung +121° (c=1 in Chloroform)
Formelmasse 506.5
Reinheit (%) 98%
Physikalische Form Pulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'