Learn More
Methyl-2-(4-Isobutylphenyl)propanoat, Thermo Scientific™
Catalog Number p-4391409
Click to view available options
Menge:
1 g
10 g
25 g
Verpackung:
Bernsteinfarbene Glasflasche
61566-34-5
C14H20O2
220.312
YNZYUHPFNYBBFF-UHFFFAOYSA-N
ibuprofen methyl ester, methyl 2-4-isobutylphenyl propanoate, motrin methyl ester, methyl 2-4-isobutylphenyl propionate, methyl 2-4-2-methylpropyl phenyl propanoate, 2-4-isobutylphenyl propionic acid methyl ester, racemic ibuprofen methyl ester, s-+-ibuprofen methyl ester, 2r/s-2-4-isobutylphenyl-propionic acid methyl ester, benzeneacetic acid,, a-methyl-4-2-methylpropyl-,methyl ester
109101
Methyl-2-[4-(2-methylpropyl)phenyl]propanoat
CC(C)CC1=CC=C(C=C1)C(C)C(=O)OC
61566-34-5
C14H20O2
220.312
YNZYUHPFNYBBFF-UHFFFAOYSA-N
ibuprofen methyl ester, methyl 2-4-isobutylphenyl propanoate, motrin methyl ester, methyl 2-4-isobutylphenyl propionate, methyl 2-4-2-methylpropyl phenyl propanoate, 2-4-isobutylphenyl propionic acid methyl ester, racemic ibuprofen methyl ester, s-+-ibuprofen methyl ester, 2r/s-2-4-isobutylphenyl-propionic acid methyl ester, benzeneacetic acid,, a-methyl-4-2-methylpropyl-,methyl ester
109101
Methyl-2-[4-(2-methylpropyl)phenyl]propanoat
CC(C)CC1=CC=C(C=C1)C(C)C(=O)OC
Methyl 2-(4-Isobutylphenyl)propanoat
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2-(4-isobutylphenyl)propanoate
Prozentgehaltsbereich 97%
Verpackung Bernsteinfarbene Glasflasche
Menge 1 g
Formelmasse 220.31
Reinheit (%) 97%