missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-acetamido-5-Chlorbenzoat, 97 %, Thermo Scientific™

Catalog Number p-7023364
Click to view available options
Menge:
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
20676-54-4
C10H10ClNO3
227.64
MFCD00144759
TVAAIYFBEWHVCV-UHFFFAOYSA-N
2-acetamido-5-chlorobenzoic acid methyl ester, methyl 5-chloro-2-acetamidobenzoate, methyl n-acetyl-5-chloroanthranilate, n-acetyl-5-chloroanthranilic acid methyl ester, 2-acetylamino-5-chloro-benzoic acid methyl ester, methyl 2-acetylamino-5-chlorobenzoate, pubchem3337, acmc-1cioq, methyl2-acetamido-5-chlorobenzoate, methyl 2-acetamido-5-chloranyl-benzoate
4640571
methyl 5-chloro-2-acetamidobenzoate
COC(=O)C1=CC(Cl)=CC=C1NC(C)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
20676-54-4
C10H10ClNO3
227.64
MFCD00144759
TVAAIYFBEWHVCV-UHFFFAOYSA-N
2-acetamido-5-chlorobenzoic acid methyl ester, methyl 5-chloro-2-acetamidobenzoate, methyl n-acetyl-5-chloroanthranilate, n-acetyl-5-chloroanthranilic acid methyl ester, 2-acetylamino-5-chloro-benzoic acid methyl ester, methyl 2-acetylamino-5-chlorobenzoate, pubchem3337, acmc-1cioq, methyl2-acetamido-5-chlorobenzoate, methyl 2-acetamido-5-chloranyl-benzoate
4640571
methyl 5-chloro-2-acetamidobenzoate
COC(=O)C1=CC(Cl)=CC=C1NC(C)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2-acetamido-5-chlorobenzoate
Schmelzpunkt 128°C to 131°C
Menge 50 g
Beilstein 2808490
Formelmasse 227.65
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'