missing translation for 'onlineSavingsMsg'
Learn More

Methyl2-formylthiophen-4-carboxylat, 95 %, Thermo Scientific Chemicals

Catalog Number p-7030063
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
67808-66-6
C7H6O3S
170.182
MFCD01859945
GNXNZRYWBFMVHK-UHFFFAOYSA-N
methyl 2-formyl-4-thiophenecarboxylate, 2-formylthiophene-4-carboxylic acid methylester, 5-formyl-3-thiophenecarboxylic acid methyl ester, 5-formyl-3-thiophene carboxylic acid methylester, pubchem7741, tpc-h014, methyl 5-methanoylthiophene-3-carboxylate, 5-formyl-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 5-formyl-, methyl ester, 3-thiophenecarboxylicacid, 5-formyl-, methyl ester
818925
Methyl5-Formylthiophen-3-Carboxylat
COC(=O)C1=CSC(=C1)C=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
67808-66-6
C7H6O3S
170.182
MFCD01859945
GNXNZRYWBFMVHK-UHFFFAOYSA-N
methyl 2-formyl-4-thiophenecarboxylate, 2-formylthiophene-4-carboxylic acid methylester, 5-formyl-3-thiophenecarboxylic acid methyl ester, 5-formyl-3-thiophene carboxylic acid methylester, pubchem7741, tpc-h014, methyl 5-methanoylthiophene-3-carboxylate, 5-formyl-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 5-formyl-, methyl ester, 3-thiophenecarboxylicacid, 5-formyl-, methyl ester
818925
Methyl5-Formylthiophen-3-Carboxylat
COC(=O)C1=CSC(=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2-formylthiophene-4-carboxylate
Schmelzpunkt 113°C to 117°C
Menge 250 mg
Empfindlichkeit Air Sensitive
Formelmasse 170.19
Reinheit (%) 95%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'