missing translation for 'onlineSavingsMsg'
Learn More

Methyl2-methyl-3-furoat, 97 %, Thermo Scientific Chemicals

Catalog Number p-7081794
Click to view available options
Menge:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6141-58-8
C7H8O3
140.138
MFCD00003249
UVRRIABXNIGUJZ-UHFFFAOYSA-N
80237
Methyl 2-Methylfuran-3-Carboxylat
CC1=C(C=CO1)C(=O)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6141-58-8
C7H8O3
140.138
MFCD00003249
UVRRIABXNIGUJZ-UHFFFAOYSA-N
80237
Methyl 2-Methylfuran-3-Carboxylat
CC1=C(C=CO1)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2-methyl-3-furoate
Dichte 1.114
Siedepunkt 71°C to 72°C (20 mmHg)
Flammpunkt 63°C (145°F)
Brechungsindex 1.4745
Menge 25 g
Beilstein 1342040
Synonym methyl 2-methyl-3-furoate, methyl 2-methyl-3-furancarboxylate, 2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 2-methyl-, methyl ester, 3-furoic acid, 2-methyl-, methyl ester, methyl2-methyl-3-furoate, acmc-1bcvo, methyl 2-methyl-3-furoate #, methyl-2-methyl-3-furoate, 2-methylfuran-3-carboxylic acid methyl
Formelmasse 140.14
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'