Learn More
Methyl-3,4-O-Isopropyliden-bL-erythro-pentopyranosid-2-ulose, TRC
Specifications
Specifications
| Chemischer Name oder Material | Methyl 3,4-O-Isopropylidene-beta-L-erythro-pentopyranosid-2-ulose |
| CAS | 4096-62-2 |
| Summenformel | C9H14O5 |
| Menge | 100 mg |
| Synonym | 3,4-O-(1-Methylethylidene)-beta-L-erythro-pentopyranosid-2-ulose, methyl 3,4-O-Isopropylidene-L-erythro-pentopyranosidulose |
| InChI-Formel | InChI=1S/C9H14O5/c1-9(2)13-5-4-12-8(11-3)6(10)7(5)14-9/h5,7-8H,4H2,1-3H3/t5-,7-,8-/m0/s1 |
| SMILES | CO[C@H]1OC[C@@H]2OC(C)(C)O[C@@H]2C1=O |
| IUPAC-Name | (3aS,6S,7aS)-6-methoxy-2,2-dimethyl-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran-7-on |
| Molekulargewicht (g/mol) | 202.2 |
| Formelmasse | 202.0841 |