missing translation for 'onlineSavingsMsg'
Learn More

Methyl3-aminocrotonat, 97 %, Thermo Scientific Chemicals

Catalog Number p-7027895
Click to view available options
Menge:
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
14205-39-1
C5H9NO2
115.13
MFCD00008072,MFCD00008072,MFCD00008072
XKORCTIIRYKLLG-ONEGZZNKSA-N
methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester
643918
methyl (2E)-3-aminobut-2-enoate
COC(=O)\C=C(/C)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
14205-39-1
C5H9NO2
115.13
MFCD00008072,MFCD00008072,MFCD00008072
XKORCTIIRYKLLG-ONEGZZNKSA-N
methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester
643918
methyl (2E)-3-aminobut-2-enoate
COC(=O)\C=C(/C)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 3-aminocrotonate
Schmelzpunkt 81°C to 85°C
Siedepunkt 122°C (42 mmHg)
Flammpunkt 91°C (195°F)
Menge 100 g
Beilstein 956592
Empfindlichkeit Hygroscopic
Formelmasse 115.13
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'