missing translation for 'onlineSavingsMsg'
Learn More

Methyl-3-(furfurylthio)-propionat, 98 %, Thermo Scientific™

Catalog Number p-7031360
Click to view available options
Menge:
1 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
94278-26-9
C9H12O3S
200.252
MFCD00053109
MXXNUXUTQGTBGJ-UHFFFAOYSA-N
methyl 3-furfurylthio propionate, unii-l1387h8y3u, methyl furfuryl mercaptopropionate, methyl 3-2-furylmethylsulfanyl propanoate, 3-furfurylthio propanoic acid methyl ester, 3-furfurylthio propionic acid methyl ester, propanoic acid, 3-2-furanylmethyl thio-, methyl ester, methyl 3-furan-2-ylmethyl sulfanyl propanoate, methyl3-furfurylthio propionate
15225386
Methyl 3-(Furan-2-ylmethylsulfanyl)Propanoat
COC(=O)CCSCC1=CC=CO1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
94278-26-9
C9H12O3S
200.252
MFCD00053109
MXXNUXUTQGTBGJ-UHFFFAOYSA-N
methyl 3-furfurylthio propionate, unii-l1387h8y3u, methyl furfuryl mercaptopropionate, methyl 3-2-furylmethylsulfanyl propanoate, 3-furfurylthio propanoic acid methyl ester, 3-furfurylthio propionic acid methyl ester, propanoic acid, 3-2-furanylmethyl thio-, methyl ester, methyl 3-furan-2-ylmethyl sulfanyl propanoate, methyl3-furfurylthio propionate
15225386
Methyl 3-(Furan-2-ylmethylsulfanyl)Propanoat
COC(=O)CCSCC1=CC=CO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 3-(furfurylthio)propionate
Dichte 1.163
Siedepunkt 114°C to 116°C (1.12 mmHg)
Flammpunkt 92°C (198°F)
Brechungsindex 1.512
Menge 1 g
Formelmasse 200.26
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'