missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetamido-5-Chlor-2-Methoxybenzoat, 98 %

Catalog Number p-7025412
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4093-31-6
C11H12ClNO4
257.67
MFCD00048191
OUEXNQRVYGYGIK-UHFFFAOYSA-N
methyl 4-acetamido-5-chloro-o-anisate, unii-3d7z5o4uxa, benzoic acid, 4-acetylamino-5-chloro-2-methoxy-, methyl ester, 3d7z5o4uxa, methyl 4-acetylamino-5-chloro-2-methoxybenzoate, methyl 5-chloro-4-acetamido-2-methoxybenzoate, 2-methoxy-4-acetylamino-5-chloro methyl benzoate, 4-acetamido-5-chloro-2-methoxybenzoic acid methyl ester, acmc-1afsv, dsstox_cid_31529
77721
COC(=O)C1=CC(Cl)=C(NC(C)=O)C=C1OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4093-31-6
C11H12ClNO4
257.67
MFCD00048191
OUEXNQRVYGYGIK-UHFFFAOYSA-N
methyl 4-acetamido-5-chloro-o-anisate, unii-3d7z5o4uxa, benzoic acid, 4-acetylamino-5-chloro-2-methoxy-, methyl ester, 3d7z5o4uxa, methyl 4-acetylamino-5-chloro-2-methoxybenzoate, methyl 5-chloro-4-acetamido-2-methoxybenzoate, 2-methoxy-4-acetylamino-5-chloro methyl benzoate, 4-acetamido-5-chloro-2-methoxybenzoic acid methyl ester, acmc-1afsv, dsstox_cid_31529
77721
COC(=O)C1=CC(Cl)=C(NC(C)=O)C=C1OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Schmelzpunkt 153°C to 156°C
Menge 10 g
IUPAC-Name Methyl-5-chlor-4-acetamido-2-methoxybenzoat
Formelmasse 257.67
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'