Learn More
Methyl-4-pyridilketoxim, 97 %, Thermo Scientific™
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Methyl 4-pyridyl ketoxime |
| Menge | 1 g |
| Synonym | 4-acetylpyridine oxime, 1-4-pyridinyl-1-ethanone oxime, methyl 4-pyridyl ketoxime, 4-1-nitrosoethylidene-1h-pyridine, n-1-pyridin-4-yl ethylidene hydroxylamine, ethanone, 1-4-pyridinyl-, oxime, z-1-pyridin-4-yl ethanone oxime, e-n-1-pyridin-4-yl ethylidene hydroxylamine, ethanone,1-4-pyridinyl-, oxime, 1e-1-4-pyridinyl ethanone oxime |
| Formelmasse | 136.15 |
| Reinheit (%) | 97% |