Learn More
Methyl-6-chlorpyridin-2-carboxylat, 95 %, Thermo Scientific Chemicals
Catalog Number p-7055442
Click to view available options
Menge:
5 g
25 g
100 g
6636-55-1
C7H6ClNO2
171.58
MFCD00155391
TWUXBVMXSBEKHA-UHFFFAOYSA-N
methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester
242723
Methyl 6-Chlorpyridin-2-Carboxylat
COC(=O)C1=NC(=CC=C1)Cl
6636-55-1
C7H6ClNO2
171.58
MFCD00155391
TWUXBVMXSBEKHA-UHFFFAOYSA-N
methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester
242723
Methyl 6-Chlorpyridin-2-Carboxylat
COC(=O)C1=NC(=CC=C1)Cl
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2?-bipyridine-6-carboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Methyl6-chlorpyridin-2-carboxylat wurde mit 2-(Trimethylstannyl)pyridin gekoppelt, um Methyl2,2′-bipyridin-6-carboxylat zu erzeugen.
Löslichkeit
Schwer in Wasser löslich.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von starken Oxidationsmitteln und Hitze fernhalten.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 6-chloropyridine-2-carboxylate
Schmelzpunkt 92°C to 98°C (literature)
Farbe Gelb
Menge 5 g
Beilstein 128580
Löslichkeitsinformationen Slightly soluble in water.
Formelmasse 171.58
Reinheit (%) 95%
Physikalische Form Fest