missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranosid, 98 %

Catalog Number 15450657
Click to view available options
Menge:
500 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
115457-83-5
C15H29NO7
335.40
MFCD00077425
XPIVOYOQXKNYHA-RGDJUOJXSA-N
hecameg, 6-o-n-heptylcarbamoyl methylglucoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxytetrahydro-2h-pyran-2-yl methyl heptylcarbamate, methyl 6-o-n-heptylcarbamoyl-a-d-glucopyranoside, methyl 6-o-n-heptylcarbamoyl-alpha-d-glucopyranoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxyoxan-2-yl methyl n-heptylcarbamate, 6-o-n-heptylcarbamoyl methyl-alpha-d-glucopyranoside, 6-o-n-heptylcarbamoyl-methyl-alpha-d-glucopyranoside, a-d-glucopyranoside,methyl,6-heptylcarbamate, 6-o-n-heptylcarbamoyl-methyl-, a-d-glucopyranoside
2733955
[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
CCCCCCCNC(=O)OC[C@H]1O[C@H](OC)[C@H](O)[C@@H](O)[C@@H]1O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
115457-83-5
C15H29NO7
335.40
MFCD00077425
XPIVOYOQXKNYHA-RGDJUOJXSA-N
hecameg, 6-o-n-heptylcarbamoyl methylglucoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxytetrahydro-2h-pyran-2-yl methyl heptylcarbamate, methyl 6-o-n-heptylcarbamoyl-a-d-glucopyranoside, methyl 6-o-n-heptylcarbamoyl-alpha-d-glucopyranoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxyoxan-2-yl methyl n-heptylcarbamate, 6-o-n-heptylcarbamoyl methyl-alpha-d-glucopyranoside, 6-o-n-heptylcarbamoyl-methyl-alpha-d-glucopyranoside, a-d-glucopyranoside,methyl,6-heptylcarbamate, 6-o-n-heptylcarbamoyl-methyl-, a-d-glucopyranoside
2733955
[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
CCCCCCCNC(=O)OC[C@H]1O[C@H](OC)[C@H](O)[C@@H](O)[C@@H]1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside
Schmelzpunkt 100°C
Farbe Weiß
Menge 500 mg
Beilstein 3560078
Merck Index 14,4621
Optische Drehung +101° (c=1 in Water)
Formelmasse 335.39
Reinheit (%) 98%
Physikalische Form Pulver
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'