missing translation for 'onlineSavingsMsg'
Learn More

Methylcellulose, Viskosität 1600 cPs

Catalog Number p-7058741
Click to view available options
Menge:
100 g
500 g
2.5 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
9004-67-5
C20H38O11
454.513
MFCD00081763
YLGXILFCIXHCMC-JHGZEJCSSA-N
51063134
(5R)-2,3,4-Trimethoxy-6-(Methoxymethyl)-5-[(2S)-3,4,5-Trimethoxy-6-(Methoxymethyl)oxan-2-yl]oxyoxan
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
9004-67-5
C20H38O11
454.513
MFCD00081763
YLGXILFCIXHCMC-JHGZEJCSSA-N
51063134
(5R)-2,3,4-Trimethoxy-6-(Methoxymethyl)-5-[(2S)-3,4,5-Trimethoxy-6-(Methoxymethyl)oxan-2-yl]oxyoxan
COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. Methyl cellulose, as a gel, has the unique property of setting when hot and melting when cold. Methyl cellulose is used as a variable viscosity personal lubricant. Methyl cellulose is used in the manufacture of capsules in nutritional supplements. Methyl cellulose finds a major application as a performance additive in construction materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl cellulose
Namenshinweis Viscosity: 1600cPs
Geruch Odorless
Menge 100 g
Merck Index 14,6040
Löslichkeitsinformationen Readily Soluble in a water below 50°C and wide variety of solvents.
Physikalische Form Pulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'