missing translation for 'onlineSavingsMsg'
Learn More

Methylperfluordodecanoat, 96 %, Thermo Scientific™

Catalog Number p-7046919
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56554-52-0
C13H3F23O2
628.128
MFCD00236616
ABNZIVVYHQYSLI-UHFFFAOYSA-N
methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
547885
Methyl2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoat
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56554-52-0
C13H3F23O2
628.128
MFCD00236616
ABNZIVVYHQYSLI-UHFFFAOYSA-N
methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
547885
Methyl2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoat
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl perfluorododecanoate
Schmelzpunkt 54°C to 56°C
Dichte 1.66
Strukturformel CF3(CF2)10CO2CH3
Menge 5 g
Formelmasse 628.13
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'