Learn More
Methyldiphenylsulfoniumtetrafluorborat, 95 %, Thermo Scientific Chemicals
Catalog Number p-7043379
Click to view available options
Menge:
5 g
25 g
10504-60-6
C13H13BF4S
288.11
MFCD00051861
LHAMVDBAOJCBDW-UHFFFAOYSA-N
diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
2758717
Methyl(Diphenyl)sulfanium;Tetrafluoroborat
F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
10504-60-6
C13H13BF4S
288.11
MFCD00051861
LHAMVDBAOJCBDW-UHFFFAOYSA-N
diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
2758717
Methyl(Diphenyl)sulfanium;Tetrafluoroborat
F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Methyldiphenylsulfoniumtetrafluoroborat ist ein Methylentransferreagenz. Es wird auch in Alkyldiphenylsulfoniumsalzen anstelle von Trialkyl verwendet, um schwefelhaltige Nebenprodukte aus der Sommelet-Hauser-Umlagerung zu vermeiden.
Löslichkeit
Löslich in Wasser, Alkohol, Chloroform.
Hinweise
Kühl und trocken in einem gut versiegelten Behälter lagern. Behälter dicht verschlossen lassen.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyldiphenylsulfonium tetrafluoroborate
Schmelzpunkt 63°C to 67°C
Strukturformel (C6H5)2SCH3BF4
Menge 5 g
UN-Nummer UN1759
Beilstein 4732019
Formelmasse 288.11
Reinheit (%) 95%