missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl3-amino-4(Methylsulfonyl)thiophol-2-carboxylat, 97 %, Thermo Scientific™
Click to view available options
Menge:
1 g
10 g
Spezifikation
Chemischer Name oder Material | Methyl 3-amino-4-(methylsulfonyl)thiophene-2-carboxylate |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 1 g |
Synonym | methyl 3-amino-4-methylsulfonyl thiophene-2-carboxylate, methyl 3-amino-4-methanesulfonylthiophene-2-carboxylate, methyl 3-amino-4-methylsulphonyl thiophene-2-carboxylate, 3-amino-4-methanesulfonyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-4-methylsulfonyl thiophene, maybridge1_004594, timtec-bb sbb005527, buttpark 33\04-59, methyl3-amino-4-methylsulfonyl thiophene-2-carboxylate, methyl 3-amino-4-methylsulfonyl thiophene-2carboxylate |
Formelmasse | 235.28 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'