missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 17629216
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17629216 100 mL 100mL
15401707 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17629216 Supplier Thermo Scientific Alfa Aesar Supplier No. 041252.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylmagnesium bromide, 3M in ether has the wide range of applications in organic synthesis. Alkylation of aldehydes and ketones is the most common applications of Grignard reagent. It is a strong nucleophile that can form new carbon-carbon bonds. Also like organolithium compounds, it is useful to form carbon-heteroatom bonds.

Solubility
Soluble in ether, toluene and tetrahydrfuran. Insoluble in hydrocarbons.

Notes
It is highly flammable and reacts with moisture or water to form methane gas, so it should be stored in tightly closed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Brown
Physical Form Liquid
Quantity 100 mL
Linear Formula CH3MgBr
UN Number UN1928
Flash Point -40°C (-40°F)
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
IUPAC Name magnesium;carbanide;bromide
Formula Weight 119.26
Concentration or Composition (by Analyte or Components) 3M in ether
Odor Ether-like
Packaging ChemSeal™ bottles
Sensitivity Air and moisture sensitive
Density 1.035 g/mL
Chemical Name or Material Methylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.