missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15073-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.Spezifikation
METTL3 | |
Polyclonal | |
Unconjugated | |
METTL3 | |
IME4, M6A, methyltransferase like 3, METTL3, MT A70, MTA70, Spo8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361035, 56335, 56339 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86U44, Q8C3P7 | |
Mettl3 | |
METTL3 Fusion Protein Ag7110 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |