Beschreibung
The mGluR1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC2144 and based on accession number NM_838. The protein contains a C-terminal DDK tag. |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens glutamate receptor, metabotropic 1 (GRM1), transcript variant 1, mRNA. |
| Wirtsspezies | Human |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Forschungskategorie | GPCR |
| Protein State | Native |
Nur für Forschungszwecke