missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGP Mouse anti-Human, Mouse, Clone: 1A1C3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60055-1-IG-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.Spezifikation
MGP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08493, P19788 | |
Mgp | |
MGP Fusion Protein Ag1091 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
1A1C3 | |
Unconjugated | |
MGP | |
GIG36, matrix Gla protein, MGLAP, MGP, NTI | |
Mouse | |
Protein A | |
RUO | |
17313, 4256 | |
-20°C | |
Liquid |