missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (MEM-E/08), mFluor Violet 450 SE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
MHC Class I Monoclonal antibody specifically detects MHC Class I in Human samples. It is validated for Flow Cytometry
Spezifikation
Spezifikation
| Antigen | MHC Class I |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | MEM-E/08 |
| Konjugat | mFluor Violet 450 SE |
| Verdünnung | Flow Cytometry |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 |
| Wirtsspezies | Mouse |
| Immunogen | Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide. |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?