missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ NovaCel™ Wischtücher

Saugfähige PVA-Wischtücher in besonders sauberer Qualität und hoher Chemikalienbeständigkeit

Marke:  Micronova™ WP20-99

Weitere Details : CAS : 000-00-0 Netto-Gewicht : 1.00000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13107346

  • 38.70 € / 100 Stück


Verfügbar ab
27-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Micronova Cleanroom Wipes include a range of low-linting, absorbent wipes ideal for cleaning, sanitizing and wiping applications in a variety of industries.

  • Cleanroom wipes for a variety of applications
  • Low-linting and absorbent properties
  • Available in a range of fabrics for specialized cleaning:
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
    • Microfiber — 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
    • PolyMesh — 100% polyester with a layer of mesh provides a mild scrubbing surface
    • NovaCel — Solid polyvinyl acetate, super absorbent, removes fine particles and dander
  • Available in a range of sizes
  • Available gamma irradiated for aseptic environments
  • Good compatibility with chemicals
  • Launderable for reuse
  • Autoclavable
Spezifikation

Spezifikation

Store at ambient temperature
White
Parts and Surface Cleaning, to Remove Tough Soils, and Liquid Removal
MegaTex, Non-Woven Polyamide
No
10/Pk.
24.1 cm
24.1 x 24.1 cm (9.5 x 9.5 in.)
24.1 cm
Cleanroom Wipes
For workstations, laminar flow benches
Non-sterile
Videos
Dokumentation