Learn More
C-Mount-Adapter zum Anschluss von Digital-Kameras, für die Steddy-Serie
Marke: Medline Scientific 7357.0965
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Trinocular configuration allows for the addition of a camera for digital imaging or video camera.The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
• Robust stereo zoom microscope
• Integral single piece column stand eliminates vibration and image distortion
• Ergonomic diamond shaped base
• Total magnification range 2.5x to 120x
• Variable intensity transmitted and incident light sources