Learn More
C-Mount-Adapter zum Anschluss von Digital-Kameras, für die Steddy-Serie
Marke:  Medline Scientific 7357.0965
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 12526687
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Trinocular configuration allows for the addition of a camera for digital imaging or video camera.
The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
• Robust stereo zoom microscope
• Integral single piece column stand eliminates vibration and image distortion
• Ergonomic diamond shaped base
• Total magnification range 2.5x to 120x
• Variable intensity transmitted and incident light sources