Learn More
Invitrogen™ Midkine Monoclonal Antibody (1011522)
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
Spezifikation
Spezifikation
| Antigen | Midkine |
| Anwendungen | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 1011522 |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with trehalose and no preservative |
| Gen | Mdk |
| Gen-Zugriffsnummer | P21741 |
| Gen-Alias | amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein |
| Mehr anzeigen |