missing translation for 'onlineSavingsMsg'
Learn More

Milnacipran hydrochloride, Tocris Bioscience™

5-HT and noradrenalin re-uptake inhibitor (SNRI)

Marke:  Tocris (Bio-Techne) 328650

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412724

  • 555.12 € / 50 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Orally active 5-HT and noradrenalin re-uptake inhibitor (SNRI) (IC 50 values are 203 and 100 nM respectively) that displays no affinity at a range of other receptors. Causes adaptive changes to α 1 -adrenergic and 5-HT 2A serotonergic systems when administered repeatedly. Exhibits antidepressive and antinociceptive activities in vivo.
Spezifikation

Chemische Identifikatoren

101152-94-7
282.812
dalcipran, ixel, rac-milnacipran hydrochloride, 1r,2s-rel-2 aminomethyl-n,n-diethyl-1-phenyl-cyclopropanecarboxamide, monohydrochloride, 2s-2-aminomethyl-n,n-diethyl-1-phenylcyclopropane-1-carboxamide hydrochloride
(2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
C15H23ClN2O
XNCDYJFPRPDERF-LKNSGBSQSA-N
45039934
CCN(CC)C(=O)C1(CC1CN)C2=CC=CC=C2.Cl

Spezifikation

Milnacipran hydrochloride
101152-94-7
dalcipran, ixel, rac-milnacipran hydrochloride, 1r,2s-rel-2 aminomethyl-n,n-diethyl-1-phenyl-cyclopropanecarboxamide, monohydrochloride, 2s-2-aminomethyl-n,n-diethyl-1-phenylcyclopropane-1-carboxamide hydrochloride
CCN(CC)C(=O)C1(CC1CN)C2=CC=CC=C2.Cl
282.812
282.81
50mg
C15H23ClN2O
XNCDYJFPRPDERF-LKNSGBSQSA-N
(2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
45039934
>99%
Videos