Learn More
ML 161, Tocris Bioscience™
Inhibits PAR1-mediated platelet activation
Marke:  RD Systems (Bio-Techne) 4570/10
Weitere Details : Netto-Gewicht : 0.00010kg
Menge. 10mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13414944
  • 224.41 € / 10 Milligramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Inhibitor of protease-activated receptor 1 (PAR1)-mediated platelet activation (IC50 = 0.26μM for the inhibition of platelet P-selectin expression on human platelets). Thought to act allosterically. Also inhibits thrombin-induced platelet activation.
Spezifikation
Chemische Identifikatoren
423735-93-7
361.239
2-bromo-n-3-butyramidophenyl benzamide, 2-bromo-n-3-butanoylamino phenyl benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl benzamide, 2-bromo-n-3-butyrylamino phenyl benzamide, cambridge id 5946339, aronis27179, benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl, ml hplc, n-3-butyrylamino phenyl-2-bromobenzamide, 2-bromo-n-3-butanamidophenyl benzamide
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
C17H17BrN2O2
DFOVLSMXPWPCFH-UHFFFAOYSA-N
1048267
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br
Spezifikation
ML 161
423735-93-7
2-bromo-n-3-butyramidophenyl benzamide, 2-bromo-n-3-butanoylamino phenyl benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl benzamide, 2-bromo-n-3-butyrylamino phenyl benzamide, cambridge id 5946339, aronis27179, benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl, ml hplc, n-3-butyrylamino phenyl-2-bromobenzamide, 2-bromo-n-3-butanamidophenyl benzamide
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br
361.239
361.23
10mg
C17H17BrN2O2
DFOVLSMXPWPCFH-UHFFFAOYSA-N
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
1048267
>98%
Videos