missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Alle ansehen Proteintech ProdukteBeschreibung
The MMP3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MMP3. This antibody recognizes human, mouse, rat antigen. The MMP3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | MMP3 |
| Anwendungen | Immunofluorescence |
| Konzentration | 1000 μg/mL |
| Konjugat | CoraLite Plus 488 |
| Gensymbole | CHDS6, Matrix metalloproteinase 3, MMP 3, MMP3, SL 1, STMY, STMY1, STR1, Stromelysin 1, Transin 1 |
| Wirtsspezies | Rabbit |
| Menge | 100 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 4314 |
| Zielspezies | Human, Mouse, Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?