Learn More
MOB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561212
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SKAKPNGKKP AAEERKAYLE PEHTKARITD FQFKELVVLP REIDLNEWLA SNTTTFFHHI NLQYSTISEF CT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
HCCA2 binds to and stimulates the kinase activity of the related human serine-threonine kinases NDR1 and NDR2.Spezifikation
MOB2 | |
Polyclonal | |
Unconjugated | |
MOB2 | |
1110017M21Rik; 2700078K21Rik; AI256456; HCCA2; Mmh; MOB kinase activator 2; Mob2; Mob2 homolog; MOB2 Mps One Binder homolog; Mps one binder kinase activator-like 2; Ovary-specific MOB-like protein; RGD1562983 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81532 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q70IA6 | |
MOB2 | |
Recombinant protein corresponding to Human MOB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only