Learn More
MOCOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561632
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NQGLLYDRSW MVVNHNGVCL SQKQEPRLCL IQPFIDLRQR IMVIKAKGME PIEVPLEENS ERTQIRQSRV CADRVSTYDC GEKISSWLST FFGRPCHLIK QSSNSQRNAK KK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.
MOCOS sulfurates the molybdenum cofactor of xanthine dehydrogenase (XDH), which is required for their enzymatic activities (Ichida et al., 2001).Spezifikation
MOCOS | |
Polyclonal | |
Unconjugated | |
MOCOS | |
1110018O12Rik; HMCS; MCS; moCo sulfurase; MOCOS; Molybdenum cofactor sulfurase; molybdenum cofactor sulfurtransferase; MOS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EN8 | |
MOCOS | |
Recombinant protein corresponding to Human MOCOS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only