Learn More
MOCS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563532
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KANLLTTEEI LTLARLFVKE GIDKIRLTGG EPLIRPDVVD IVAQLQRLEG LRTIGVTTNG INLARLLPQL QKAGLSAIN Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
MOCS1 Polyclonal antibody specifically detects MOCS1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
MOCS1 | |
Polyclonal | |
Unconjugated | |
MOCS1 | |
3110045D15Rik; AI043170; AW536397; bHLHe36; Cell migration-inducing gene 11 protein; Cyclic pyranopterin monophosphate synthase; GTP 3',8-cyclase; MIG11; migration-inducing gene 11 protein; MIR; Mlx interactor; MOCOD; MOCODA; MOCS1; MOCS1A enzyme; molybdenum cofactor biosynthesis protein 1; Molybdenum cofactor biosynthesis protein A; Molybdenum cofactor biosynthesis protein C; molybdenum cofactor synthesis 1; molybdenum cofactor synthesis-step 1 protein A-B; molybdenum cofactor synthesis-step 1 proteins A and B; MONDOA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZB8 | |
MOCS1 | |
Recombinant protein corresponding to Human MOCS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only