missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ C2 Overall, Modell CHZ5
Click to view available options
Größe:
Medium
Large
X-Large
2X-Large
3X-Large
Spezifikation
Produkttyp | Hooded Coveralls |
Größe | Large |
Material | Tychem |
Farbe | Yellow |
Taillenform | Elastic |
Knöchelform | Elastic |
Stulpenform | Double |
Flaschenhalstyp | Hooded |
Hood Type | Elastic |
Verschlusstyp | Zipper with Storm Flap |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'