missing translation for 'onlineSavingsMsg'
Learn More

MOG Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA554531

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15762461

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Immunogen sequence: ALVGDEVELP CRISPGKNAT GMEVGWYRPP FSRVVHLYRN GKDQDGDQAP EYRGRTELLK DAIGEGKVTL RIRNVRFSDE GGFTCFFRDH SYQEEAAMEL KVEDPFYWVS PG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Myelin / Oligodendrocyte Specific Protein (MOSP) (pI of ∽6.7) is found exclusively on oligodendrocytes and myelin in the central nervous system (CNS). Myelin is produced by oligodendrocytes in the CNS and Schwann cells in the peripheral nervous system (PNS). Defining the differences between CNS and PNS myelin may provide insights into the process of myelination and the pathogenesis of certain idiopathic diseases involving myelin such as multiple sclerosis, certain leukodystrophies, and other myelinopathies. On demyelination, immunoreactivity for MOSP, like MBP, is lost.
Spezifikation

Spezifikation

MOG
Polyclonal
Unconjugated
MOG
MOG; MGC26137; myelin oligodendrocyte glycoprotein; DAQB-92E24.2; MOGIG-2; MOG Ig-AluB; MOG alpha-6
Rabbit
Antigen affinity chromatography
RUO
4340
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q16653
MOG
Recombit protein corresponding to Human MOG
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only