Learn More
Monomethyl auristatin E (MMAE) Antibody (08), Novus Biologicals™
Artikelnummer. 30115289 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
20 μg
Packungsgröße:
100 Mikrogramm
20 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30115289 20 μg 20 Mikrogramm
30116704 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30115289 Lieferant Novus Biologicals Lieferanten-Nr. NBP33197420ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
Monomethyl auristatin E (MMAE) Monoclonal antibody specifically detects Monomethyl auristatin E (MMAE) in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Monomethyl auristatin E (MMAE)
Anwendungen ELISA
Klassifikation Monoclonal
Klon 08
Konjugat Unconjugated
Verdünnung ELISA 1:1000-1:2000
Zusammensetzung 0.2 um filtered solution in PBS
Gen-Alias 474645-27-7, Monomethyl auristatin E (GMP), vedotin
Wirtsspezies Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin E (MMAE) conjugated to BSA
Reinigungsverfahren Protein A purified
Menge 20 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Non-species specific
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1