Learn More
CD25 Mouse anti-Human, BUV563, Clone: 2A3, BD
Mouse Monoclonal Antibody
276.84 € - 586.50 €
Spezifikation
Antigen | CD25 |
---|---|
Klon | 2A3 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Brilliant Ultraviolet 563 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15804479
|
BD Biosciences
565700 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
15894469
|
BD Biosciences
565699 |
100 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
The 2A3 monoclonal antibody specifically binds to human CD25, the low-affinity alpha subunit of the Interleukin-2 Receptor (IL- 2Rα). CD25 associates with CD122 (IL-2Rβ chain) and CD132 (common γchain or γc) to form the high-affinity signal-transducing IL -2R complex. CD25 is expressed by subsets of thymocytes and peripheral blood lymphocytes including CD4+CD25+ regulatory T cells and memory T cells. CD25 antigen density increases on activated T cells including phytohemagglutinin (PHA)-, concanavalin A (Con A)-, and CD3-activated T lymphocytes. High levels of CD25 can be expressed by T lymphocytes from mixed lymphocyte cultures and by human T-lymphocyte leukemia virus (HTLV)-infected T-lymphocyte leukemia lines, for example, HUT-102. CD25 can also be expressed by activated B cells and macrophages. Recombinant IL-2 blocks the binding of the 2A3 antibody to PHA-activated T lymphocytes.
The antibody was conjugated to BD Horizon BUV563 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 which has an Ex Max of 348nm and an acceptor dye. The tandem has an Em Max at 563 nm. BD Horizon BUV563 can be excited by the 355nm ultraviolet laser. On instruments with a 561nm Yellow-Green laser, the recommended bandpass filter is 585/15nm with a 535nm long pass to minimize laser light leakage. When BD Horizon BUV563 is used with an instrument that does not have a 561nm laser, a 560/40nm filter with a 535nm long pass may be more optimal. Due to the excitation and emission characteristics of the acceptor dye, there may be spillover into the PE and PE-CF594 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Spezifikation
CD25 | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Phytohemagglutinin-activated T Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2A3 | |
Monoclonal | |
Mouse | |
Human | |
100 | |
IgG1 κ | |
Affinity Purified | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 |
For Research Use Only.