Learn More
Mouse anti-Human IgE Heavy Chain Secondary Antibody (E454 (5D5)), PE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
IgE Heavy Chain Monoclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for ELISA
Spezifikation
Spezifikation
| Antigen | IgE Heavy Chain |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | E454 (5D5) |
| Konjugat | PE |
| Zusammensetzung | PBS |
| Gen-Alias | Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon |
| Wirtsspezies | Mouse |
| Immunogen | Immunoglobulin E from human serum |
| Reinigungsverfahren | Protein G purified |
| Mehr anzeigen |