missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sh3gl1, Mouse, APC, Clone: S51-1, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Sh3gl1.
Marke: Abnova MAB11882.100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
Sh3gl1 | |
Monoclonal | |
APC | |
Western Blot (1 ug/mL) Immunocytochemistry (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) Immunohistochemistry (0.1-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Sh3gl1 | |
Sh3gl1 | |
A synthetic peptide corresponding to amino acids of 287-302 of rat Sh3gl1. | |
100 μg | |
Primary | |
Rat | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
S51-1 | |
Mouse monoclonal antibody raised against synthetic peptide of Sh3gl1. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
SH3P8 | |
Mouse | |
Protein G Purification | |
RUO | |
81922 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |