missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC4, Mouse, Polyclonal Antibody, Abnova™
Beschreibung
Sequence: EITSLDTENIDEILNNADVALVNFYADWCRFSQMLHPIFEEASDVIKEEFPNENQVVFARVDCDQHSDIAQRYRISKYPTLKLFRNGMMMKREYRGQRSVKALADYIRQQKSDPIQEIRDLAEITTLDRSKRNIIGYFEQKDSDNYRVFERVANILHDDCAFLSAFGDVSKPERYSGDNIIYKPPGHSAPDMVYLGAMTNFDVTYNWIQDKCVPLVREITFENGEELTEEGLPFLILFHMKEDTESLEIFQNEVARQLISEKGTINFLHADCDKFRHPLLHIQKTPADCPVIAIDSFRHMYVFGDFKDVLIPGKLKQFVFDLHSGKLHREFHHGPDPTDTAPGEQAQDVASSPPESSFQKLAPSEYRYTLLRDRDEL
Spezifikation
Spezifikation
| Antigen | TXNDC4 |
| Anwendungen | ELISA |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Beschreibung | Mouse polyclonal antibody raised against a full-length recombinant TXNDC4. |
| Zusammensetzung | 50% glycerol |
| Gen | TXNDC4 |
| Gen-Zugriffsnummer | BC005374 |
| Gen-Alias | ERP44/KIAA0573 |
| Gensymbole | TXNDC4 |
| Mehr anzeigen |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?