missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse B7-H5 / Gi24 / VISTA Protein, His Tag
423.38 € - 2661.24 €
Spezifikation
Zugriffsnummer | NP_001153044.1 |
---|---|
Molekulargewicht | 19.7 kDa |
Quelle | Mouse B7-H5, His Tag (B75-M52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 191 (Accession # Q9D659-1). |
Immunogen | Phe 33 - Ala 191 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15885977
|
ACROBiosystems
B75-M52H7-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15895977
|
ACROBiosystems
B75-M52H7-1MG |
1 mg |
2661.24 €
1 Milligramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Platelet receptor Gi24, also known as B7-H5 and stress-induced secreted protein-1 (Sisp-1), is a protein that in humans is encoded by the C10orf54 gene, which contains 1 Ig-like (immunoglobulin-like) domain. As for C10orf54 gene, C10orf54 appears to positively interact with BMP-4, potentiating BMP signaling and the transition from an undifferentiated to a differentiated state on ESCs. Human C10orf54 undergoes proteolytic cleavage by MT1-MMP, generating a soluble 30 kDa extracellular fragment plus a 25-30 kDa membrane-bound fragment.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.7kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001153044.1 | |
Mouse B7-H5, His Tag (B75-M52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 191 (Accession # Q9D659-1). | |
-20°C | |
B7-H5 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
19.7 kDa | |
Phe 33 - Ala 191 | |
B7-H5,SISP1,Gi24,VISTA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |